Learn More
Enzo Life Sciences Grp 75 Mortalin (human), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP828F
Encompass_Preferred
Alternative name: MtHsp70, HspA9, Heat shock 70 kDa protein 9. MW: ~75kDa. Purity: ≥80% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 3% glycerol and 150mM sodium chloride. Source: Produced in E. coli. UniProt: P38646. Application: WB. Recommended dilutions: Western Blot (100ng, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Long Term Storage: -80°C
Catalog No. 50-946-993