Learn More
Enzo Life Sciences Grp78/BiP ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002140001
Encompass_Preferred
High-sensitivity ELISA kit for quantifying both normal and upregulated levels of Grp78/BiP for unfolded protein response, cancer and neurodegenerative disease research. The Grp78/BiP ELISA kit is a colorimetric, competitive immunoassay kit with results in 2.5 hours. Alternative Name: Glucose-regulated protein 78. Binding immunoglobulin protein. Sensitivity: 8.4ng/ml (range 1.4-4500ng/ml). Assay Time: 2.5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter Plate, Conjugate, Antibody, Assay Buffer, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, Extraction Reagent
Catalog No. NC1277557