Learn More
Enzo Life Sciences Grp 78 BiP (hamster), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP765D
Encompass_Preferred
Alternative name: HspA5, Hsp70-5, Heat shock 70 kDa protein 5. Purity: ≥80% (SDS-PAGE; Western blot). MW: ~78kDa. Formulation: Liquid. In 50mM Tris, pH 7.5, containing 150mM NaCl, 10% glycerol and 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7638