Learn More
Enzo Life Sciences Grp 78 BiP (hamster), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP765F
Encompass_Preferred
Alternative name: HspA5, Hsp70-5, Heat shock 70 kDa protein 5. MW: ~78kDa. Purity: ≥80% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM Tris, pH 7.5, containing 150mM NaCl, 10% glycerol and 1mM DTT. Source: Produced in E. coli. UniProt: P07823. Application: WB. Long Term Storage: -80°C
Catalog No. NC1276769