Learn More
Enzo Life Sciences Grp78 BiP polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA7690200
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of rat Grp78 (BiP). The sequence is completely conserved in mouse and hamster. Source: Purified from rabbit serum. Species reactivity: Mouse, Rat, Xenopus, Rabbit, Monkey, Hamster, Fungus, Bovine. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7468