Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7184RTM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM249. Clonality: Monoclonal. Host: Rat. Isotype: IgG2a kappa. Positive Control: U20S cells. About 50% of breast tumors express grp94 protein. Cellular Staining: Cytoplasmic, nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified glucose regulated protein 94 (grp94) from chicken oviducts. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 94kDa.Chromosome: 12q23.3. Gene Symbol: HSP90B1.
Catalog No. 50-188-5244