Learn More
Enzo Life Sciences Grp 94 monoclonal antibody (9G10) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA850F
Encompass_Preferred
Clone: 9G10. Host: Rat. Isotype: IgG2a. Immunogen: Native chicken Grp94. UniProt: P08110. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: ICC, IF, IHC (PS), IP, WB, Electron microscopy. Recommended dilutions: Immunoprecipitation (1:100). Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-645