Learn More
Enzo Life Sciences Grp94 (canine), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP766F
Encompass_Preferred
Alternative name: Tra1, Tumor rejection antigen 1, Hsp90B1, Gp96, Heat shock protein 90 kDa beta member 1, Endoplasmin. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~94kDa. Formulation: Liquid. In 20mM MOPS, pH 7.5, containing 0.5mM EDTA. Source: Produced in Sf21 insect cells. Produced in a baculovirus expression system. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7640