Learn More
Enzo Life Sciences Grp 94 monoclonal antibody (9G10) (DyLight 488 conjugate) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA850488D
Encompass_Preferred
Clone: 9G10. Host: Rat. Isotype: IgG2a. Immunogen: Native chicken Grp94. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide.
Catalog No. 50-200-7562