Learn More
Enzo Life Sciences GrpE (E. coli) polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA240J
Encompass_Preferred
Host: Rabbit. Immunogen: Purified E. coli GrpE. UniProt ID: P09372 (strain K12). Source: Purified from rabbit serum. Species reactivity: E. coli. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7427