Learn More
Enzo Life Sciences GrpE (E. coli), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP650F
Encompass_Preferred
Alternative name: HSP70 cofactor, Nucleotide exchange factor, HSP24, Heat shock protein 24. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~24kDa. Formulation: Liquid. In 25mM HEPES, pH 7.6, containing 50mM potassium chloride and 5.0% glycerol. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB in vitro Assay.
Catalog No. 50-200-7618