Learn More
Aviva Systems Biology Corp GRPEL1 Antibody - N-terminal region (ARP46346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRPEL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRPEL1. Alternate names: FLJ25609, HMGEApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-014-59308