Learn More
Medchemexpress LLC Grpp (human) | 1132745-52-8 | 98.9% | 3384.47 | 500 UG
SDP
Catalog No. 5000398136
Encompass_Preferred
GRPP (human) is a 30 amino acid Gcg-derived peptide that causes slight increases in plasma insulin and decreases in plasma glucagon. It does not affect insulin secretion in rat islets.
  • 30 amino acid Gcg-derived peptide
  • Causes slight increases in plasma insulin
  • Decreases plasma glucagon
  • Does not affect insulin secretion in rat islets
  • Requires sealed storage, away from moisture
  • Powder stable for 2 years at -80°C
  • Powder stable for 1 year at -20°C
  • In solvent stable for 6 months at -80°C
  • In solvent stable for 1 month at -20°C
Catalog No. 50-003-98136 Supplier Medchemexpress LLC Supplier No. HYP5074500UG
GRPP (human) is a 30 amino acid Gcg-derived peptide that causes slight increases in plasma insulin and decreases in plasma glucagon. It does not affect insulin secretion in rat islets.
  • 30 amino acid Gcg-derived peptide
  • Causes slight increases in plasma insulin
  • Decreases plasma glucagon
  • Does not affect insulin secretion in rat islets
  • Requires sealed storage, away from moisture
  • Powder stable for 2 years at -80°C
  • Powder stable for 1 year at -20°C
  • In solvent stable for 6 months at -80°C
  • In solvent stable for 1 month at -20°C