Learn More
Aviva Systems Biology Corp GRSF1 Antibody - N-terminal region (ARP40381_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40381_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GRSF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRSF1. Alternate names: ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-57420