Learn More
Strem, An Ascensus Company CAS 172222-30-9. 25g. Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium 97% [Grubbs Catalyst Gen. 1].
SDP
Supplier:  Strem, An Ascensus Company 44094425G
Encompass
CAS 172222-30-9. 25g. Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium 97% [Grubbs Catalyst Gen. 1]. . Molecular Weight 822.97. Molecular Formula C43H72C2P2Ru. Color/form purple pwdr. Strem 44-0944. http//www.strem.com/catalog/v/44-0944/
Catalog No. 50-211-0832