Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Grubbs Catalyst™ M102

Catalog No. 57972650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-972-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-972-650G Supplier Sigma Aldrich Supplier No. 57972650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Grubbs Catalyst™ M102

Specifications

CAS 172222-30-9
Melting Point 153°C
Molecular Formula C43H72Cl2P2Ru
Linear Formula C43H72Cl2P2Ru
Quantity 50 g
Synonym Benzylidene-bis(tricyclohexylphosphine)dichlororuthenium; Bis(tricyclohexylphosphine)benzylidine ruthenium(IV) dichloride; Dichloro(benzylidene)bis(tricyclohexylphosphine)ruthenium(II); Grubbs Catalyst(R) 1st Generation; Grubbs Catalyst(R) M1a (C823)
Molecular Weight (g/mol) 822.96 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.