Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Grubbs Catalyst™ M103

Catalog No. 9086141G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-861-41G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 90-861-41G Supplier Sigma Aldrich Supplier No. 9086141G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Umicore Grubbs Catalyst M103 is a homogeneous catalyst useful for alkene metathesis, cross metathesis, ring-closing metathesis, and self metathesis.Product of UmicoreAdditional information available

Specifications

CAS 915797-21-6
Melting Point 200°C to 204°C
Molecular Formula C24H22O3PRh
Linear Formula C24H22O3PRh
Quantity 1 g
Synonym ROPAC
Molecular Weight (g/mol) 492.31 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.