Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Grubbs Catalyst™ M300

Catalog No. 682330500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-233-0500MG 500 mg
68-233-02G 2 g
68-233-0100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 68-233-0500MG Supplier Sigma Aldrich Supplier No. 682330500MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Grubbs Catalyst™ M300

Specifications

CAS 900169-53-1
Melting Point 140°C
Molecular Formula C38H40Br2Cl2N4Ru
Linear Formula C38H40Br2Cl2N4Ru
MDL Number MFCD09265164
Quantity 500 mg
Synonym Grubbs Catalyst(R) 3rd Generation; Grubbs Catalyst(R) M37a (C884); Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](benzylidene)bis(3-bromopyridine)ruthenium(II); Grubbs Catalyst(R) C884; [1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(phenylmethylene)bis(3-bromopyridine)ruthenium(II); [1,3-Dimesityl-2-imidazolidinylidene]dichloro(phenylmethylene)bis(3-bromopyridine)ruthenium(II)
Molecular Weight (g/mol) 884.54 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.