Learn More
Vee Gee Vee Gee Specific Gravity Hydrometer, 0.900 to 1.000, 0.001 subdivisions, 305 mm
Catalog No. NC2748915
Encompass_Preferred
The Precision Specific Gravity Hydrometer is calibrated for use at 60F (15.55C). Specific Gravity Range: 0.900 to 1.000 Subdivisions: 0.001 Approx. Length: 305 mm
Catalog No. NC2748915 Supplier Vee Gee Supplier No. 660312
The Precision Specific Gravity Hydrometer is calibrated for use at 60F (15.55C). Specific Gravity Range: 0.900 to 1.000 Subdivisions: 0.001 Approx. Length: 305 mm