Learn More
Abcam GS 39783, GABAB positive allosteric modulator, 50MG
Catalog No. 502707680
Encompass_Preferred
MW 337.4 Da, Purity >99%. Novel, GABAB positive allosteric modulator (EC50 = 3 μM for [35S]GTPγS binding). Shows anxiolytic and antiaddictive effects *in vivo.* Blood-brain barrier permeable*.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7680 Supplier Abcam Supplier No. AB12032050MG
MW 337.4 Da, Purity >99%. Novel, GABAB positive allosteric modulator (EC50 = 3 μM for [35S]GTPγS binding). Shows anxiolytic and antiaddictive effects *in vivo.* Blood-brain barrier permeable*.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement