Learn More
Medchemexpress LLC HY-103586 10mg Medchemexpress, GS-441524 CAS:1191237-69-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10358610MG
Encompass_Preferred
Medchemexpress, HY-103586 10mg GS-441524 CAS:1191237-69-0 GS-441524 could strongly inhibits feline infectious peritonitis virus (FIPV), with an EC50 of 0.78 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8130