Learn More
Medchemexpress LLC GS-443902 trisodium | 1355050-21-3 | 10MG
SDP
Catalog No. 5000235546
Encompass_Preferred
GS-443902 trisodium (GS-441524 triphosphate trisodium) is a potent viral RNA-dependent RNA-polymerases (RdRp) inhibitor with IC50s of 1 1 M 5 M for RSV RdRp and HCV RdRp respectively GS-443902 trisodium is the active triphosphate metabolite of Remdesivir (GS-5734)[1][2]
Catalog No. 50-002-35546 Supplier Medchemexpress LLC Supplier No. HY126303C10MG
GS-443902 trisodium (GS-441524 triphosphate trisodium) is a potent viral RNA-dependent RNA-polymerases (RdRp) inhibitor with IC50s of 1 1 M 5 M for RSV RdRp and HCV RdRp respectively GS-443902 trisodium is the active triphosphate metabolite of Remdesivir (GS-5734)[1][2]