Learn More
Medchemexpress LLC GS-443902 trisodium (GS-441524 triphosphate trisodium) | 1355050-21-3 | 100.0% | 597.15 g/mol | C12H13N5Na3O13P3 | 25MG
SDP
Supplier:  Medchemexpress LLC HY126303C25MG
Encompass_Preferred
GS-443902 trisodium is the triphosphate metabolite of remdesivir and a potent inhibitor of viral RNA-dependent RNA polymerases (RdRp). It is used in antiviral research to study polymerase inhibition and remdesivir pharmacology, and is supplied as a high-purity, water-soluble trisodium salt suitable for in vitro and in vivo formulation studies.
  • Active triphosphate metabolite of remdesivir
  • Inhibits viral RNA-dependent RNA polymerases
  • High reported purity for analytical work
  • Water soluble; requires ultrasonic for full dissolution
  • Suitable for in vitro and in vivo formulation studies
  • Requires low-temperature storage and light protection
Catalog No. 50-002-36660