Learn More
TARGETMOL CHEMICALS INC GS-9191 5MG
SDP
Catalog No. 502580750
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GS-9191 is a potent inhibitor of DNA polymerase alpha and ß a prodrug of the novel nucleoside analogue 9-(2-phosphonomethoxyethyl)guanine (PMEG) which penetrates the skin and is metabolized in the epithelium to an active nucleoside triphosphate analogue.GS-9191 has antiproliferative activity and reduces the size of papillomas in a dose-related manner.GS-9191 may be useful in the treatment of HPV-induced lesions. purity: 98%
Catalog No. 50-258-0750 Supplier TARGETMOL CHEMICALS INC Supplier No. T254665MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GS-9191 is a potent inhibitor of DNA polymerase alpha and ß a prodrug of the novel nucleoside analogue 9-(2-phosphonomethoxyethyl)guanine (PMEG) which penetrates the skin and is metabolized in the epithelium to an active nucleoside triphosphate analogue.GS-9191 has antiproliferative activity and reduces the size of papillomas in a dose-related manner.GS-9191 may be useful in the treatment of HPV-induced lesions. purity: 98%