Learn More
TARGETMOL CHEMICALS INC GS-9667 5MG
SDP
Catalog No. 502580613
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GS-9667 a selective and partial agonist of the A(1) adenosine receptor (AR) represents an effective therapy for Type 2 diabetes (T2DM) and dyslipidemia via lowering of free fatty acids (FFA). purity: 99%
Catalog No. 50-258-0613 Supplier TARGETMOL CHEMICALS INC Supplier No. T274395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GS-9667 a selective and partial agonist of the A(1) adenosine receptor (AR) represents an effective therapy for Type 2 diabetes (T2DM) and dyslipidemia via lowering of free fatty acids (FFA). purity: 99%