Learn More
Montana Molecular Gs Mutant Expression Kit - BacMam, 30mL - Montana Molecular
SDP
Catalog No. NC3787574
Encompass_Preferred
A BacMam vector expressing a constitutively active mutant Gs protein. As BacMam is readily titratable, this kit enables you to fine-tune intracellular cAMP levels without the use of chemicals like forskolin. It can be co-expressed with our cADDis cAMP sensors, especially when investigating Gi signaling.
Catalog No. NC3787574 Supplier Montana Molecular Supplier No. X0250X030
A BacMam vector expressing a constitutively active mutant Gs protein. As BacMam is readily titratable, this kit enables you to fine-tune intracellular cAMP levels without the use of chemicals like forskolin. It can be co-expressed with our cADDis cAMP sensors, especially when investigating Gi signaling.