Learn More
GUARDIAN EQUIPMENT Replacement spray head covers for all Guardian Eyewash units. Each head has a “flip top” dust cover, internal flow control and filter to remove impurities from the water flow.
Catalog No. NC2941320
Encompass_Preferred
Replacement spray head covers for all Guardian Eyewash units. Each head has a “flip top” dust cover, internal flow control and filter to remove impurities from the water flow.
Catalog No. NC2941320 Supplier GUARDIAN EQUIPMENT Supplier No. AP470002ORGR
Replacement spray head covers for all Guardian Eyewash units. Each head has a “flip top” dust cover, internal flow control and filter to remove impurities from the water flow.