Learn More
GUARDIAN EQUIPMENT GS-Plus™ Spray Head
Catalog No. NC0872518
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Spray Head for Guardian Eyewash and WideArea™ Eye/Face Wash Units. Features integral flip top dust cover and 1.6 GPM flow control.
Catalog No. NC0872518 Supplier GUARDIAN EQUIPMENT Supplier No. AP470001
This item has a minimum qty of 3 per supplier requirements.
Spray Head for Guardian Eyewash and WideArea™ Eye/Face Wash Units. Features integral flip top dust cover and 1.6 GPM flow control.