Learn More
Medchemexpress LLC GS143 | 916232-21-8 | MFCD30182334 | 99.1% | 466.46 g/mol | C28H19FN2O4 | 5 MG
SDP
Catalog No. 5000263300
Encompass_Preferred
GS143 is a small-molecule inhibitor of IκBα ubiquitination used for laboratory research. It inhibits SCFβTrCP1-mediated ubiquitination of IκBα (IC50 5.2 μM) and suppresses TNFα-induced NF-κB signaling. Available in solid and solution formats, it is supplied for research use only.
Catalog No. 50-002-63300 Supplier Medchemexpress LLC Supplier No. HY1102615MG
GS143 is a small-molecule inhibitor of IκBα ubiquitination used for laboratory research. It inhibits SCFβTrCP1-mediated ubiquitination of IκBα (IC50 5.2 μM) and suppresses TNFα-induced NF-κB signaling. Available in solid and solution formats, it is supplied for research use only.