Learn More
Enzo Life Sciences GS 28 monoclonal antibody (HFD9) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPT047D
Encompass_Preferred
Clone: HFD9. Host: Mouse. Isotype: IgG1. Immunogen: Rat liver Golgi-enriched membranes. UniProt: Q62931. Source: Purified from ascites. Species Reactivity: Mouse, Rat, Sheep, Rabbit, Porcine, Hamster, Dog, Chicken, Bovine. Applications: WB, Electron microscopy. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-838