Learn More
Enzo Life Sciences GS28 monoclonal antibody (HFD9) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPT047F
Encompass_Preferred
Clone: HFD9. Host: Mouse. Isotype: IgG1. Immunogen: Rat liver Golgi-enriched membranes. Source: Purified from ascites. Species reactivity: Mouse, Rat, Sheep, Rabbit, Porcine, Hamster, Dog, Chicken, Bovine. Applications: WB Electron microscopy. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7690