Learn More
TARGETMOL CHEMICALS INC GSA-10 5MG
SDP
Catalog No. 502580919
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. GSA-10 is a potent Smoothened (Smo) receptor agonist with an EC50 of 1.2 uM. It acts on Smo to promote the differentiation of pluripotent mesenchymal stem cells into osteoblasts. It also mediates Hedgehog (Hh) signaling which may have research implications for regenerative medicine in cancer diseases. purity: 98%
Catalog No. 50-258-0919 Supplier TARGETMOL CHEMICALS INC Supplier No. T228145MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. GSA-10 is a potent Smoothened (Smo) receptor agonist with an EC50 of 1.2 uM. It acts on Smo to promote the differentiation of pluripotent mesenchymal stem cells into osteoblasts. It also mediates Hedgehog (Hh) signaling which may have research implications for regenerative medicine in cancer diseases. purity: 98%