Learn More
STEMCELL Technologies GSA 10, Size: 50 mg
SDP
Supplier:  STEMCELL Technologies 73174
Encompass
GSA 10 is an agonist of Smoothened (SMO), a cell surface receptor and mediator of the hedgehog signaling pathway. It is a quinolinecarboxamide derivative that binds in a distinct binding pocket from cyclopamine. GSA10 is active at an EC₅₀ of 1.2 μM in an alkaline phosphatase-based mouse mesenchymal cell line (C3H10T1/2) differentiation assay (Gorojankina et al.).DIFFERENTIATION· Promotes differentiation of multipotent mesenchymal progenitor cells into osteoblasts (Gorojankina et al.).
Catalog No. 50-197-6037