Learn More
Aviva Systems Biology Corp GSC Antibody - N-terminal region (ARP32174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GSC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GSC. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-54474