Learn More
GSC Go Science Crazy Beaker Tongs - 10 in. Plastic Coated
SCI_ED
Catalog No. S13744 Shop All GSC Products
Encompass
Encompass_Preferred
Not a toy. Product intended for ages 13 and up.
Catalog No. S13744 Supplier GSC Go Science Crazy Supplier No. 4BT7210
Not a toy. Product intended for ages 13 and up.
A must have for the lab when using beakers that are too hot to handle. The GSC Beaker Tongs - 10 in. Plastic Coated are used for handling beakers with a 50-2000ml capacity. Measure 10 in. long and have a firm but gentle grip.
  • Holds 50-2000ml beakers
  • Firm but soft grip
  • Plastic coated
Specifications
Length (English) 10 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov