Learn More
GSC Go Science Crazy Single Burette Clamp
SCI_ED
Catalog No. S13742 Shop All GSC Products
Click to view available options
Coating:
Plastic
Uncoated
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating
S13742 Uncoated
S13743 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up.
Catalog No. S13742 Supplier GSC Go Science Crazy Supplier No. 4BC37UC15
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up.
A clamp specifically designed to use with burettes. The GSC Burette Clamp Single with Jaws is a cast-aluminum clamp with nickel-plated steel thumb-screw.
  • Cast aluminum clamp
  • Nickel-plated steel thumb screw
  • Jaw range of 3/8 in. to 1-3/8 in.
Specifications
Type Burette Clamp
Material Cast Aluminum, Nickel-plated Steel
For Use With (Equipment) For use with burettes, support stands, rods
For Use With (Application) For use with GSC Burettes, Support Stands and Rods
Coating Uncoated
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov