Learn More
GSC Go Science Crazy Density Rod
SCI_ED
Catalog No. S13642 Shop All GSC Products
Click to view available options
Includes:
Set of 12 Aluminum
Catalog No. Includes
S13642 Set of 12 Aluminum
1 options
Catalog No. S13642 Supplier GSC Go Science Crazy Supplier No. DNRDSTAL
To compare the density of different materials
  • Set of 12 aluminum rods
  • Comes in a square wood block
  • Explore mass, density and volume
Specifications
Product Type Mass density volume
Includes Set of 12 Aluminum
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov