Learn More
GSC Go Science Crazy Electrode Carbon with Stopper
SCI_ED
Catalog No. S13613 Shop All GSC Products
Click to view available options
Material:
Carbon, Rubber
Catalog No. Material
S13613 Carbon, Rubber
1 options
Not a toy. Product intended for ages 13 and up. May Contain Latex!
Catalog No. S13613 Supplier GSC Go Science Crazy Supplier No. 4501045
Not a toy. Product intended for ages 13 and up. May Contain Latex!
A carbon electrode to be used in conjunction with the GSC Hoffman Electrolysis. The GSC Electrode Carbon with Stopper is a carbon rod that can be fitted into a rubber stopper, to be used with a Hoffman Electrolysis.
  • 0.25 in. diameter
  • Carbon rod
  • Fitted into rubber stopper
  • For use with Hoffman Electrolysis
Specifications
Material Carbon, Rubber
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov