Learn More
GSC Go Science Crazy Form Truss Boom
Catalog No. S13618 Shop All GSC Products
Change view
Click to view available options
Form truss, boom
Catalog No.
S13618 Form truss, boom
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13618 Supplier GSC Go Science Crazy Supplier No. FRCTRO1
Not a toy. Product intended for ages 13 and up.
Includes
Half meter ruler (50cm), 2 Knife edge clamps (fully adjustable), Support rod clamp
A great tool for helping students understand how bridges are engineered. The GSC Form Truss Boom can be used to help students understand the Law of Composition of Force. The apparatus also aids students in understanding concepts such as compression and tension. The Form Truss Boom is a great tool for helping students understand how bridges are engineered.
  • Shows Law of Composition of Force
  • 2 Knife edge clamps
  • Support rod clamp
Order Info
Rod, base, spring scale, and mass set sold separately