Learn More
GSC Go Science Crazy Hooke's Law Demonstration
SCI_ED
Catalog No. S13640 Shop All GSC Products
Click to view available options
For Use With (Application):
Demonstrates Hooke's Law
Catalog No. For Use With (Application)
S13640 Demonstrates Hooke's Law
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13640 Supplier GSC Go Science Crazy Supplier No. 480810
Not a toy. Product intended for ages 13 and up.
A great classroom apparatus to help students understand the principles of Hooke's Law. The GSC Hooke's Law Demonstration includes a sturdy base, testing spring, metal scale, an indicator needle, and a complete set of hooked weights. Instructions included.
  • Effectively demonstrates Hooke's Law
  • Includes everything needed
  • Comes with instructions
Specifications
Product Type Hooke's Law Apparatus
For Use With (Application) Demonstrates Hooke's Law
Includes Sturdy base and rod; Testing spring; Metal scale; Indicator needle; Set of slotted weights (one 100 g hanger with a red pointer, nine 50 g weights)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov