Learn More
GSC Go Science Crazy Incline Plane Friction Set
SCI_ED
Catalog No. S13858 Shop All GSC Products
Click to view available options
Incline plane, friction set
Catalog No.
S13858 Incline plane, friction set
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13858 Supplier GSC Go Science Crazy Supplier No. IPFRIC811
Not a toy. Product intended for ages 13 and up.
Includes
Two scales, aluminum wheels and cone bearings to minimize friction, aluminum weight, hanger (50 grams), instruction manual.
A great addition to any science classroom, the Incline Plane Friction Set demonstrates the resolution of forces, uniform and accelerated motion, and potential energies. The GSC Incline Plane Friction Set can be used for multiple demonstrations including the resolution of forces, uniform and accelerated motion, and potential energies.
  • Used for multiple demonstrations
  • Can be set between 0 and 45 degrees
  • Instructions included
Specifications
Product Type Incline Plane Friction Set
For Use With (Application) Demonstrations of resolution of forces, uniform and accelerated motion, and potential energies
Includes 2 scales; 3 boards (two wood, one aluminum); 1 weight hanger
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov