Learn More
GSC Go Science Crazy Convection of Liquid Glass Square
SCI_ED
Catalog No. S41803A Shop All GSC Products
Change view
Click to view available options
Convection of liquid square
Catalog No.
S41803A Convection of liquid square
1 options
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up.
Catalog No. S41803A Supplier GSC Go Science Crazy Supplier No. 30310
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Not a toy. Product intended for ages 13 and up.
Demonstrates properties of convection currents. The water and dye filled glass frame is heated at the bottom corner. The convection currents are seen as the die is moved through he glass tube.
  • Used for Convection of Liquid
  • Made of borosilicate glass
Specifications
Product Type Convection of liquid glass
For Use With (Application) Convection of Liquid
Subject Physics