Learn More
GSC Go Science Crazy Electrostatic Charge Kit
SCI_ED
Catalog No. S09587 Shop All GSC Products
Click to view available options
For Use With (Application):
Experimenting with static electricity
Catalog No. For Use With (Application)
S09587 Experimenting with static electricity
1 options
Catalog No. S09587 Supplier GSC Go Science Crazy Supplier No. ELSTCH01
Includes
Electrophorus with 15cm acrylic charge plate and insulating handle, six different friction rods, acetate cloth, polyethylene film, proof plane with 15mm ball, neon lamps, graphite ball with hook, 12 pith balls, and instruction manual.
Allows students to make electricity by friction and to store and transfer electricity.
Specifications
Product Type Electrostatic Charge Kit
Includes 1 Wool cloth; 1 Silk cloth; 1 Cotton cloth; 1 Acetate cloth; 1 Neon lamp; 6 Pairs of Pith Balls; 6 Friction rods (acrylic, glass, polyethylene, polystyrene, PVC, nylon); 1 Acrylic rod with a hook; 1 Conductive ball with a hook; 1 Spool of string; 1 Balloon; 1 Electrophorus and charge plate
For Use With (Application) Experimenting with static electricity
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov