Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Galvanometers
SCI_ED

Catalog No. S77886A Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Product Type:
Galvanometer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S77886A Galvanometer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Not a toy. Product intended for ages 13 and up.

Catalog No. S77886A Supplier GSC Go Science Crazy Supplier No. 4130920
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Not a toy. Product intended for ages 13 and up.

For electrical experiments in lab or classroom.

  • Full scale and fully enclosed
  • Range of -500 to 0 to +500μA
  • Accuracy of ±2.5%
  • Color-coded binding posts
  • Can be used with banana cords

Specifications

Product Type Galvanometer
Description Full scale and fully enclosed
For Use With (Application) Electrical experiments in lab and classroom
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.