Learn More
GSC Go Science Crazy Hand Held DC Generator
SCI_ED
Catalog No. S13682 Shop All GSC Products
Click to view available options
Generator, hand held, dc
Catalog No.
S13682 Generator, hand held, dc
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13682 Supplier GSC Go Science Crazy Supplier No. 4185310
Not a toy. Product intended for ages 13 and up.
A great addition to any classroom experiment using up to 12 volts of DC current. The GSC Hand Held DC Generator is a simple and useful generator for experiments requiring up to 12 volts DC current. Housed in a tough plastic case with binding posts for electrical connections. Includes 2 alligator connector cords and replacement bulb.
  • Hand Held
  • Portable and easy to use
  • Posts for electrical connections
Specifications
Electrical Requirements 12 V DC
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov