Learn More
GSC Go Science Crazy Thermal Conductivity Demonstration Tool
SCI_ED
Catalog No. S08002 Shop All GSC Products
Click to view available options
Includes:
Four metal bars and support stand
Catalog No. Includes
S08002 Four metal bars and support stand
1 options
Catalog No. S08002 Supplier GSC Go Science Crazy Supplier No. THCND01
Quickly and easily compare the thermal conductivity of different metals. The GSC International Thermal Conductivity Demonstration Tool has temperature indicators marked at 1cm intervals to chart the temperature movement.
  • Quickly and easily compare the thermal conductivity of different metals
  • Four metals--copper, aluminum, brass and steel--are held in a molded support
  • Liquid Crystal temperature indicators are attached to the front of all four metal bars
  • The temperature indicators are marked at 1cm intervals to facilitate charting of the temperature movement
Specifications
Includes Four metal bars and support stand
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Not a toy. Product intended for ages 13 and up.