Learn More
GSC Go Science Crazy International Vacuum Pumps
SCI_ED
Catalog No. S13826 Shop All GSC Products
Click to view available options
Includes:
Gauge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S13826 -
S13825 Gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S13826 Supplier GSC Go Science Crazy Supplier No. 42062
Remove moisture and air from tanks and other containers with hand held vacuum pump.
  • Removes moisture and air
  • Gauge reads inches and centimeters
  • 15ml per stroke
Specifications
Pump Type Vacuum Pump
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov