Learn More
GSC Go Science Crazy Water Turbine with Dynamo Electromagnet
SCI_ED
Catalog No. S09594 Shop All GSC Products
Change view
Click to view available options
Water Turbine with Dynamo Electromagnet
Catalog No.
S09594 Water Turbine with Dynamo Electromagnet
1 options
Catalog No. S09594 Supplier GSC Go Science Crazy Supplier No. WTD01
The water turbine is used to drive a dynamo which produces electricity generating up to 6V DC that can be used for various applications.
  • Unit consists of a plastic impeller housed in a water resistant case with a clear perspex front so you can view the operation of the turbine
  • The inlet and outlet tube are provided for water to flow through the turbine
  • A dynamo is mounted along the impeller shaft that is used to light an LED, which indicates that electrical energy has been produced
Specifications
Product Type Water Turbine With Dynamo
For Use With (Application) Generating electricity
Quantity 1
Weight (English) 2 lb.
Output Power 6V DC