Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Wood Splints
SCI_ED

Catalog No. 144101A Shop All GSC Products
Change view
Click to view available options
Wood splint
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
144101A Wood splint
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 144101A Supplier GSC Go Science Crazy Supplier No. 2500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Demonstrate the presence of oxygen and supports combustion. GSC International Wood Splints are a high-quality product for use in the lab and classroom.

  • Applicator stick and planting marker
  • Thin fine wood stick
  • Approximately 5.5 L x 0.25 in. W

Specifications

Product Type Wood Splints
For Use With (Application) Demonstrate presence of Oxygen and supports combustion
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.