Learn More
GSC Go Science Crazy Linear Expansion Apparatus with Dial Indicator & 4 Metal Rods
SCI_ED
Catalog No. S13856 Shop All GSC Products
Click to view available options
Material:
Copper, Brass, Aluminum, Steel
Catalog No. Material
S13856 Copper, Brass, Aluminum, Steel
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S13856 Supplier GSC Go Science Crazy Supplier No. 1206
Not a toy. Product intended for ages 13 and up.
Includes
Dial Indicator, Steel Rod, Aluminum Rod, Brass Rod, Steel Rod
An effective tool for demonstrating the coefficient of the expansion of metals. The rod is placed inside a tube that has both an inlet and outlet for steam.
  • Shows the expansion of metals
  • Includes dial indicator and 4 rods
Specifications
Material Copper, Brass, Aluminum, Steel
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov